ClearKlens Solve VH21 Detergent
Katalog # DIVY101103248
Leverantör: Diversey
Specifikationer
- Beskrivning:ClearKlens solve VH21 detergent
- Sterility:Non sterile
- pH:<11
- Förp.:1 Bottle
Specifikationer
Om denna produkt
ClearKlens solve is an alkaline foam detergent for use in the pharmaceutical and cosmetic industries.
- Balanced blend of anionic and non-ionic surfactants in alkaline solution
- Removes oils, fats and grease of vegetable and animal origin and other product residues found in the pharmaceutical and cosmetic industries
- ClearKlens solve is for manual cleaning of process equipment, floors and walls and foam applications
- Highly effective alkaline detergent for difficult cleaning tasks
- Can be used on aluminum with care as directed
Use ClearKlens Solve for daily routine cleaning 1 to 2% v/v up to 60 °C, contact time 10 to 15 mins with brushing, followed by rinsing in clean water. Foam applications 3 to 5% v/v. For heavy duty residues requiring repeat treatments up to 10% v/v concentration may be required. ClearKlens Solve containing solutions should be thoroughly rinsed after use to remove them from all product contact surfaces.