VOLCANO Multirisk Trousers
Supplier: ALSICO
Certificates
About this item
Comfortable multirisk trousers in inherent flame retardant fabric.
- Preformed, modern fit
- Elasticated waistband with button hole
- Closure with jeans button and zipped fly
- 2 large, straight inserted side pockets
- Thigh pocket with expansion slit and phone pocket, closed with flap and hidden press studs
- Double metre pocket with flap
- 2 large back pockets with expansion slit, reinforced and closed with flap and hidden press studs
- Preformed, closable knee pockets in main fabric
- Extendable trouser legs
- Piping in accent colour
Industrial launderable.
Certifications: Oeko-Tex® standard 100; EN 1149-5 2018; IEC 61482 - 2: 2018 - APC1; EN ISO 11612:2015 (A1, B1, C1, D0, E0, F1); EN 11611:2015 - Class 1 A1; EN 13034 +A1.
Specifications
- Catalog No:
- ALSW1030940-BE52
- ALSW1030940-BE54
- ALSW1030940-BE56
- ALSW1030939-BE40
- ALSW1030940-BE58
- ALSW1030939-BE42
- ALSW1030939-BE38
- ALSW1030940-BE46
- ALSW1030940-BE48
- ALSW1030940-BE50
- ALSW1030940-BE38
- ALSW1030940-BE40
- ALSW1030940-BE42
- ALSW1030940-BE44
- ALSW1030939-BE60
- ALSW1030939-BE62
- ALSW1030939-BE64
- ALSW1030939-BE52
- ALSW1030939-BE54
- ALSW1030939-BE56
- ALSW1030939-BE58
- ALSW1030940-BE60
- ALSW1030939-BE44
- ALSW1030939-BE46
- ALSW1030940-BE62
- ALSW1030939-BE48
- ALSW1030940-BE64
- ALSW1030939-BE50
- Outfit applikationer:
- Chemical protection, Fire protection, Welding, Industrial, Mechanical protection
- Chemical protection, Fire protection, Welding, Industrial, Mechanical protection
- Chemical protection, Fire protection, Welding, Industrial, Mechanical protection
- Chemical protection, Fire protection, Welding, Industrial, Mechanical protection
- Chemical protection, Fire protection, Welding, Industrial, Mechanical protection
- Chemical protection, Fire protection, Welding, Industrial, Mechanical protection
- Chemical protection, Fire protection, Welding, Industrial, Mechanical protection
- Chemical protection, Fire protection, Welding, Industrial, Mechanical protection
- Chemical protection, Fire protection, Welding, Industrial, Mechanical protection
- Chemical protection, Fire protection, Welding, Industrial, Mechanical protection
- Chemical protection, Fire protection, Welding, Industrial, Mechanical protection
- Chemical protection, Fire protection, Welding, Industrial, Mechanical protection
- Chemical protection, Fire protection, Welding, Industrial, Mechanical protection
- Chemical protection, Fire protection, Welding, Industrial, Mechanical protection
- Chemical protection, Fire protection, Welding, Industrial, Mechanical protection
- Chemical protection, Fire protection, Welding, Industrial, Mechanical protection
- Chemical protection, Fire protection, Welding, Industrial, Mechanical protection
- Chemical protection, Fire protection, Welding, Industrial, Mechanical protection
- Chemical protection, Fire protection, Welding, Industrial, Mechanical protection
- Chemical protection, Fire protection, Welding, Industrial, Mechanical protection
- Chemical protection, Fire protection, Welding, Industrial, Mechanical protection
- Chemical protection, Fire protection, Welding, Industrial, Mechanical protection
- Chemical protection, Fire protection, Welding, Industrial, Mechanical protection
- Chemical protection, Fire protection, Welding, Industrial, Mechanical protection
- Chemical protection, Fire protection, Welding, Industrial, Mechanical protection
- Chemical protection, Fire protection, Welding, Industrial, Mechanical protection
- Chemical protection, Fire protection, Welding, Industrial, Mechanical protection
- Chemical protection, Fire protection, Welding, Industrial, Mechanical protection
- Risk area:
- Flame-retardant, Anti-static
- Flame-retardant, Anti-static
- Flame-retardant, Anti-static
- Flame-retardant, Anti-static
- Flame-retardant, Anti-static
- Flame-retardant, Anti-static
- Flame-retardant, Anti-static
- Flame-retardant, Anti-static
- Flame-retardant, Anti-static
- Flame-retardant, Anti-static
- Flame-retardant, Anti-static
- Flame-retardant, Anti-static
- Flame-retardant, Anti-static
- Flame-retardant, Anti-static
- Flame-retardant, Anti-static
- Flame-retardant, Anti-static
- Flame-retardant, Anti-static
- Flame-retardant, Anti-static
- Flame-retardant, Anti-static
- Flame-retardant, Anti-static
- Flame-retardant, Anti-static
- Flame-retardant, Anti-static
- Flame-retardant, Anti-static
- Flame-retardant, Anti-static
- Flame-retardant, Anti-static
- Flame-retardant, Anti-static
- Flame-retardant, Anti-static
- Flame-retardant, Anti-static
- Kön:
- För män
- För män
- För män
- För män
- För män
- För män
- För män
- För män
- För män
- För män
- För män
- För män
- För män
- För män
- För män
- För män
- För män
- För män
- För män
- För män
- För män
- För män
- För män
- För män
- För män
- För män
- För män
- För män
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Stängningsstorlek:
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Med dragkedja
- Tyg:
- 79% cotton FR/20% polyester/1% carbon
- 79% cotton FR/20% polyester/1% carbon
- 79% cotton FR/20% polyester/1% carbon
- 54% modacrylic/44% cotton/2% carbon
- 79% cotton FR/20% polyester/1% carbon
- 54% modacrylic/44% cotton/2% carbon
- 54% modacrylic/44% cotton/2% carbon
- 79% cotton FR/20% polyester/1% carbon
- 79% cotton FR/20% polyester/1% carbon
- 79% cotton FR/20% polyester/1% carbon
- 79% cotton FR/20% polyester/1% carbon
- 79% cotton FR/20% polyester/1% carbon
- 79% cotton FR/20% polyester/1% carbon
- 79% cotton FR/20% polyester/1% carbon
- 54% modacrylic/44% cotton/2% carbon
- 54% modacrylic/44% cotton/2% carbon
- 54% modacrylic/44% cotton/2% carbon
- 54% modacrylic/44% cotton/2% carbon
- 54% modacrylic/44% cotton/2% carbon
- 54% modacrylic/44% cotton/2% carbon
- 54% modacrylic/44% cotton/2% carbon
- 79% cotton FR/20% polyester/1% carbon
- 54% modacrylic/44% cotton/2% carbon
- 54% modacrylic/44% cotton/2% carbon
- 79% cotton FR/20% polyester/1% carbon
- 54% modacrylic/44% cotton/2% carbon
- 79% cotton FR/20% polyester/1% carbon
- 54% modacrylic/44% cotton/2% carbon
- Vikt:
- 260 g/m²
- 260 g/m²
- 260 g/m²
- 320 g/m²
- 260 g/m²
- 320 g/m²
- 320 g/m²
- 260 g/m²
- 260 g/m²
- 260 g/m²
- 260 g/m²
- 260 g/m²
- 260 g/m²
- 260 g/m²
- 320 g/m²
- 320 g/m²
- 320 g/m²
- 320 g/m²
- 320 g/m²
- 320 g/m²
- 320 g/m²
- 260 g/m²
- 320 g/m²
- 320 g/m²
- 260 g/m²
- 320 g/m²
- 260 g/m²
- 320 g/m²