Elevated Temperature Front Helmet Cover
Katalog # MMMAFC1-AL
Supplier: 3M
Specifications
- Färg:Silver
- Beskrivning:Elevated temperature front cover, aluminum
- Förp.:1
Specifications
About this item
The elevated temperature front cover is a silver aluminium front cover designed to fit on to 3M™ hard hats.
- Provide additional protection from sparks and metal splatter
- Streamlined design prevents interference with other equipment during use
- Fabric parts can be easily replaced when needed
Get extra protection from sparks and metal splatter on the front of a hard hat with the elevated temperature front cover. This silver aluminium hard hat cover has a streamlined design that helps prevent interference with other equipment during use. It fits on to 3M™ hard hats for added protection and to help reduce the possibility of particulate dropping into the users face when a faceshield is lifted. The fabric parts can be easily replaced when needed.