Scotch-Brite™ 770 Cellulosic Sponge Laminate
Katalog # MMMASCH770
Leverantör: 3M
Specifikationer
- Färg:Blå
- Beskrivning:Scotch-Brite™ Cellulosic sponge laminate, 770
- L×B:131×88 mm
- Förp.:10
Specifikationer
Om denna produkt
Scotch-Brite™ Cellulosic sponge laminate 770 is a dual-function cleaning tool made from a combination of a non-woven tablecloth containing very light abrasive particles to a pure cellulose sponge.
- Great cleaning performance
- High absorption power
- Does not scratch even the most sensitive surfaces
- Works with hot water or diluted bleach
- Excellent hydrophilic properties
Remove grease and food without leaving scratches with Scotch-Brite™ Cellulosic Sponge Laminate 770. Made from a combination of a non-woven tablecloth containing very light abrasive particles to a pure cellulose sponge. The fine texture of the synthetic fibres on the cloth makes it a medium buffer abrasive that that works with hot water or diluted bleach to remove grease and food without scratching.