Om denna produkt
Address lab’s regular- to medium-throughput blood processing needs with the Thermo Scientific™ HAEMAFlex™ six swinging bucket rotor with 6×550 ml blood bag capacity or, utilising the same bucket system, upgrade to 8×550 ml bags with the Thermo Scientific™ HAEMAFlex™ eight swinging bucket rotor.
- Eco-Spin™ technology provides energy savings up to 64% based on an engineering evaluation of windshielded and non-windshielded designs of the same rotor body
- Stainless steel rotor body, aluminum windshield and bucket, polypropylene bottle
High-throughput sample processing with the flexibility to match your current and future needs with 6 or 8×550 ml blood bag (6 or 8×1000 ml) capacity, both available with the same bucket selections. Upgrade to Thermo Scientific™ Dura-Coat™ buckets with nickel-coating to improve protection against corrosion due to moisture, chemicals or alkaline solutions.
Compatible with Thermo Scientific™ Auto-Lid™ and Auto-ID™ functions enabling instant rotor lid opening, storage and rotor identification to save time and protects the integrity of your samples.
Specifikationer
- Catalog No:
- HERA75003861
- HERA75003881
- Kapacitet:
- 6×550 ml blood bags (6×1000 ml)
- 8×550 ml blood bags (8×1000 ml)
- Rotor type:
- Swinging bucket rotor
- Swinging bucket rotor
- Max. RCF:
- 7295×g
- 7144×g
- Max. speed:
- 5000 min⁻¹
- 4600 min⁻¹