Order Entry
Sweden
ContactUsLinkComponent
Quinoline yellow sulphonic acid sodium salt (Acid yellow 3)
Irriterande
Quinoline yellow sulphonic acid sodium salt (Acid yellow 3)
Katalog # SPCMDC200-25GM
Leverantör:  Spectrum Chemical
CAS-nummer:  
Irriterande
Quinoline yellow sulphonic acid sodium salt (Acid yellow 3)
Katalog # SPCMDC200-25GM
Leverantör:  Spectrum Chemical
CAS-nummer:  

Specifikationer

  • Förpackningstyp:
    Glass, amber
  • Förp.:
    25 mg
  • Seq:
    2315

Specifikationer

Specifikationstestresultat

  • Arsenic (As):
    To Pass Test
  • Lead (Pb):
    To Pass Test

Specifikationstestresultat

Om denna produkt

 :
 :GHS07
Förvaringstemperatur:Omgivande
CAS-nummer:8004-92-0
Formel:C₁₈H₁₁NO₂.xNaO₃S
MDL-nummer:MFCD00080727