Order Entry
Sweden
ContactUsLinkComponent
Anti-P2RX7 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Anti-P2RX7 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
  BOSSBS-12106R-A750
 :  Bioss
Anti-P2RX7 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
  BOSSBS-12106R-A750
 :  Bioss

 

  • Antikroppstyp:
    Primär
  • Antikroppsnamn:
    P2RX7
  • Klonalitet:
    Polyclonal
  • Konjugation:
    Alexa Fluor® 750
  • Värd:
    Rabbit
  • ImmunKemi:
    Yes
  • ImmunoFluorescens:
    Yes
  • Isotyp:
    IgG
  • Reaktivitet:
    Human,
    Mouse
  • Western blot:
    Yes
  • Epitop:
    115-160/595
  • Form:
    Liquid
  • Gen-ID:
    5027
  • Antigen synonymer:
    P2Z receptor|P2X7|P2X purinoceptor 7|Purinergic receptor|ATP receptor|P2RX7
  • Modifiering:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molekylvikt:
    69 kDa
  • Förvaringstemperatur:
    Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Koncentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human P2RX7
  • Tested applications:
    ICC
  • Rening:
    Purified by Protein A
  • Förpackningstyp:
    Vial
  • Förp.:
    100 µl

 

 

Receptor for ATP that acts as a ligand-gated ion channel. Responsible for ATP-dependent lysis of macrophages through the formation of membrane pores permeable to large molecules. Could function in both fast synaptic transmission and the ATP-mediated lysis of antigen-presenting cells.

Type: Primary
Antigen: P2RX7
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 115-160/595
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse