Order Entry
Sweden
ContactUsLinkComponent
Anti-SLC5A3/SMIT Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Anti-SLC5A3/SMIT Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Katalog # BOSSBS-11954R-A680
Leverantör:  Bioss
Anti-SLC5A3/SMIT Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Katalog # BOSSBS-11954R-A680
Leverantör:  Bioss

Specifikationer

  • Antikroppstyp:
    Primär
  • Antikroppsnamn:
    SLC5A3/SMIT
  • Klonalitet:
    Polyclonal
  • Konjugation:
    Alexa Fluor® 680
  • Värd:
    Rabbit
  • ImmunKemi:
    Yes
  • ImmunoFluorescens:
    Yes
  • Isotyp:
    IgG
  • Reaktivitet:
    Human
  • Epitop:
    251-350/718
  • Form:
    Liquid
  • Antigen synonymer:
    Na+/myo-inositol cotransporter|SC5A3_HUMAN|SLC5A3|Solute carrier family 5 member 3|Na+/myo inositol cotransporter|solute carrier family 5 inositol transporters, member 3|SMIT2|Sodium/myo inositol cotransporter|SMIT|sodium/myo inositol cotransporter 1|Sodium/myo-inositol cotransporter
  • Modifiering:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molekylvikt:
    80 kDa
  • Förvaringstemperatur:
    Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Koncentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human SLC5A3/SMIT
  • Tested applications:
    ICC
  • Rening:
    Purified by Protein A
  • Förpackningstyp:
    Vial
  • Förp.:
    100 µl

Specifikationer

Om denna produkt

Myo-inositol is involved in many important aspects of cellular regulation including membrane structure, signal transduction and osmoregulation. It is taken up into cells by the sodium/myo-inositol cotransporter (SMIT). SMIT activity maintains intracellular concentrations of myo-inositol; it is upregulated in response to hypertonic stress. The human SMIT protein is encoded by the SLC5A3 gene, which maps to chromosome 21q22.12. It is expressed in many human tissues, such as brain, kidney and placenta. Specifically, SMIT is abundantly expressed throughout the whole brain and spinal cord in fetal rat, but is downregulated in adult rat brain with the exception of the choroid plexus, where SMIT expression remains high. In kidney, SMIT localizes to the baso-lateral membranes of the thick ascending limb of Henle (TAL) and the inner medullary collecting duct (IMCD). Impaired SMIT activity is implicated in the pathogenesis of diabetes and Down syndrome.

Type: Primary
Antigen: SLC5A3/SMIT
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 251-350/718
Host: Rabbit
Isotype: IgG
Reactivity: Human