Order Entry
Sweden
ContactUsLinkComponent
Anti-RHOG Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Anti-RHOG Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Katalog # BOSSBS-10466R-A680
Leverantör:  Bioss
Anti-RHOG Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Katalog # BOSSBS-10466R-A680
Leverantör:  Bioss

Specifikationer

  • Antikroppstyp:
    Primär
  • Antikroppsnamn:
    RHOG
  • Klonalitet:
    Polyclonal
  • Konjugation:
    Alexa Fluor® 680
  • Flödescytometri:
    Yes
  • Värd:
    Rabbit
  • ImmunKemi:
    Yes
  • ImmunoFluorescens:
    Yes
  • Isotyp:
    IgG
  • Reaktivitet:
    Human
  • Epitop:
    61-150/191
  • Form:
    Liquid
  • Gen-ID:
    391
  • Antigen synonymer:
    RHOG|ARHG|Rho-related GTP-binding protein RhoG
  • Modifiering:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molekylvikt:
    21 kDa
  • Förvaringstemperatur:
    Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Koncentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human RHOG
  • Tested applications:
    ICC
  • Rening:
    Purified by Protein A
  • Förpackningstyp:
    Vial
  • Förp.:
    100 µl

Specifikationer

Om denna produkt

Required for the formation of membrane ruffles during macropinocytosis. Plays a role in cell migration and is required for the formation of cup-like structures during trans-endothelial migration of leukocytes. In case of Salmonella enterica infection, activated by SopB and ARHGEF26/SGEF, which induces cytoskeleton rearrangements and promotes bacterial entry.

Type: Primary
Antigen: RHOG
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 61-150/191
Host: Rabbit
Isotype: IgG
Reactivity: Human