Order Entry
Sweden
ContactUsLinkComponent
Anti-GSTA3 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Anti-GSTA3 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Katalog # BOSSBS-5163R-A750
Leverantör:  Bioss
Anti-GSTA3 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Katalog # BOSSBS-5163R-A750
Leverantör:  Bioss

Specifikationer

  • Antikroppstyp:
    Primär
  • Antikroppsnamn:
    GSTA3
  • Klonalitet:
    Polyclonal
  • Konjugation:
    Alexa Fluor® 750
  • Värd:
    Rabbit
  • ImmunKemi:
    Yes
  • ImmunoFluorescens:
    Yes
  • Isotyp:
    IgG
  • Reaktivitet:
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitop:
    131-222/222
  • Form:
    Liquid
  • Gen-ID:
    2940
  • Antigen synonymer:
    GTA3|GSTA3|GSTA3-3|GST class-alpha member 3|Glutathione S-transferase A3|Glutathione S-transferase A3-3
  • Modifiering:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molekylvikt:
    25 kDa
  • Förvaringstemperatur:
    Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Koncentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human GSTA3
  • Tested applications:
    ICC
  • Rening:
    Purified by Protein A
  • Förpackningstyp:
    Vial
  • Förp.:
    100 µl

Specifikationer

Om denna produkt

Conjugation of reduced glutathione to a wide number of exogenous and endogenous hydrophobic electrophiles. catalyses isomerization reactions that contribute to the biosynthesis of steroid hormones. Efficiently catalyse obligatory double-bond isomerizations of delta(5)-androstene-3,17-dione and delta(5)-pregnene-3,2-dione, precursors to testosterone and progesterone, respectively.

Type: Primary
Antigen: GSTA3
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 131-222/222
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Rat