Orders LinkContactUsLinkComponent
Corning® 100-Layer CellCube® Module, Corning
Corning® 100-Layer CellCube® Module, Corning
Leverantör:   Corning
Corning® 100-Layer CellCube® Module, Corning
Leverantör:  Corning
Certificates
Corning® 100-Layer CellCube® Module, Corning
2 Alternativ tillgängliga nedan
Product Details & Documents
The CellCube Module is an integral, encapsulated, sterile, single-use device that is 100% pressure-hold tested before shipment.
It is comprised of a series of parallel, styrene plates joined to create thin, sealed, laminar flow spaces between adjacent plates. Each culture plate receives proprietary tissue culture treatment prior to assembly to ensure dense, uniform cell growth. These CellCube Modules provide: a series of parallel, polystyrene plates joined to create thin, sealed, laminar flow spaces between adjacent plates, 85 000cm² polystyrene growth surface optimally treated for the growth of attachment dependent cells, a perfused environment which more closely simulates in vivo conditions and reliably distributes nutrients and oxygen, an integral, encapsulated, sterile, single-use device that is 100% pressure-hold tested before shipment. Square Flask Shape.
Specifications for Product Family
Specifikationer
  • Catalog No:
  • CORN3032
  • CORN3244
  • Medievolym:
  • 6000 ml
  • 6000 ml
  • Yta:
  • Tissue culture treated
  • Tissue culture treated
Specifikationer
Product Family Options
Produktinformation
  • Odlingsyta
    Tillgänglighet
    Price
  • CORN3032
    100-layer CellCube® module, polycarbonate/polystyrene, sterile
    100-layer CellCube® module, polycarbonate/polystyrene, sterile
    85000 cm²
    Log in för att kolla tillgänglighet och ert pris
    Log in för att kolla tillgänglighet och ert pris
    1 * 1 Items
    1 * 1 Items - 50 025,00 kr
    50 025,00 kr
    1 * 1 Items
       50 025,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    CORN3032
    Medievolym
    6000 ml
    Yta
    Tissue culture treated
  • CORN3244
    100-layer CellCube® module with silicone tubing and AseptiQuik™ G, polycarbonate/polystyrene, sterile
    100-layer CellCube® module with silicone tubing and AseptiQuik™ G, polycarbonate/polystyrene, sterile
    85000 cm²
    Log in för att kolla tillgänglighet och ert pris
    Log in för att kolla tillgänglighet och ert pris
    1 * 1 Items
    1 * 1 Items - 39 025,00 kr
    39 025,00 kr
    1 * 1 Items
       39 025,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    CORN3244
    Medievolym
    6000 ml
    Yta
    Tissue culture treated