Orders LinkContactUsLinkComponent
TapeStation System, 4200
TapeStation System, 4200
4200 TapeStation system, Including laptop PC, software, vortexer, accessories, plastic consumables, user information and I&F, does not include ScreenTape consumables and reagents, I&F included, PM (preventive maintainance) not included, to be ordered separately
Katalog # 732-3992
Leverantör:   AGILENT
TapeStation System, 4200
Katalog # 732-3992
Leverantör:  AGILENT
TapeStation System, 4200
TapeStation System, 4200
4200 TapeStation system, Including laptop PC, software, vortexer, accessories, plastic consumables, user information and I&F, does not include ScreenTape consumables and reagents, I&F included, PM (preventive maintainance) not included, to be ordered separately
Price
Fråga efter pris
Log in för att kolla tillgänglighet och ert pris
1 * 1 Items
Produktdetaljer och dokument
DNA and RNA sample quality control with flexible throughput (1 to 96) at a constant cost per sample.
  • Simplify your workflow with fully automated sample processing and ready-to-use ScreenTape technology
  • Any sample number between 1 and 96 can be analysed, with constant costs per sample providing scalable throughput
  • Easily switch between DNA and RNA ScreenTape assays for excellent flexibility
  • Results are obtained quickly in as little as 1 to 2 minutes per sample, or less than 90 minutes for 96 samples
  • Achieve user-independent results with minimal manual intervention and excellent reproducibility for sizing, concentration and integrity assessment
  • Requires as little as 1 to 2 µl of DNA or RNA samples, even for high sensitivity analysis
  • Rely on the integrity standards for RNA (RNA integrity number equivalent, RINe) and genomic DNA (DNA Integrity Number, DIN)
  • Carryover is eliminated as the ScreenTape device analyses each sample in a separate lane with individual loading tips for each
  • Easy self-maintenance tasks guarantee lowest cost of ownership and maximise instrument uptime and productivity
The agilent 4200 TapeStation system is an established automated electrophoresis tool for DNA and RNA sample quality control. Fully automated sample processing enables the unattended analysis of size, concentration and integrity. Choose any sample number between 1 and 96 and analyse at constant cost per sample.
The TapeStation system provides a complete solution for true end-to-end sample quality control within any next-generation sequencing (NGS) or biobank workflow. This covers the full DNA and RNA range and offers high levels of flexibility in a simplified workflow with ready-to-use ScreenTapes.
Caution: For research use only. Not for use in diagnostic procedures.
Specifikationer
  • Beskrivning:
    4200 TapeStation system
  • Details:
    Including laptop PC, software, vortexer, accessories, plastic consumables, user information and I&F, does not include ScreenTape consumables and reagents, I&F included, PM (preventive maintainance) not included, to be ordered separately
Accessories
Produktinformation
  • Katalog #
    Details
    Tillgänglighet
    Price
  • 32-pin electrode cartridge
    32-pin electrode cartridge
    AGLS5067-5786
    For use with 4150 TapeStation (G2992A) and new 4200 TapeStation instruments (G2991B) only
    Log in för att kolla tillgänglighet och ert pris
    1 * 1 Items - 3 350,12 kr
Produktinformation
  • Katalog #
    Details
    Tillgänglighet
    Price
  • USB cable, male A - male B
    USB cable, male A - male B
    AGLS5188-8047
    For use with Agilent 4150 and 4200 TapeStation systems
    Log in för att kolla tillgänglighet och ert pris
    1 * 1 Items - 330,00 kr
Produktinformation
  • Katalog #
    Details
    Tillgänglighet
    Price
  • ScreenTape rack
    ScreenTape rack
    AGLSG2991-40007
    For use with Agilent 4200 TapeStation system
    Log in för att kolla tillgänglighet och ert pris
    1 * 1 Items - 561,18 kr
Produktinformation
  • Katalog #
    Details
    Tillgänglighet
    Price
  • Tube strip holder
    Tube strip holder
    AGLSG2992-40042
    For use with Agilent 4150 and 4200 TapeStation systems
    Log in för att kolla tillgänglighet och ert pris
    1 * 1 Items - 374,82 kr
Produktinformation
  • Katalog #
    Details
    Tillgänglighet
    Price
  • Tip waste bucket
    Tip waste bucket
    AGLSG2992-40046
    For use with Agilent 4150 and 4200 TapeStation systems
    Log in för att kolla tillgänglighet och ert pris
    1 * 1 Items - 474,35 kr
Produktinformation
  • Katalog #
    Details
    Tillgänglighet
    Price
  • Loading tips, 1 pack
    Loading tips, 1 pack
    732-3993
    For use with Agilent 4150 and 4200 TapeStation systems, 1 package contains 112 tips
    Log in för att kolla tillgänglighet och ert pris
    1 * 112 Items - 448,94 kr
Produktinformation
  • Katalog #
    Details
    Tillgänglighet
    Price
  • Loading tips, 10 pack
    Loading tips, 10 pack
    732-3994
    For use with Agilent 4150 and 4200 TapeStation systems, 1 package contains 112 tips
    Log in för att kolla tillgänglighet och ert pris
    1 * 10 Items - 4 106,12 kr
Produktinformation
  • Katalog #
    Details
    Tillgänglighet
    Price
  • Needle change cartridge
    Needle change cartridge
    AGLS5067-5783
    For use with Agilent 4150 and 4200 TapeStation systems
    Log in för att kolla tillgänglighet och ert pris
    1 * 1 Items - 2 346,35 kr