Specifications
- B×D×H, utv:747×769×1818 mm
- Kapacitet:571/419 L (gross/net)
- Beskrivning:Refrigerator FRFvg 5501
- Seq:03
- Modell:FRFvg 5501
- Elektrisk:220 - 240 V~, 2,0 A, 50 Hz
- Sound level:49 dB(A)
- Refrigerant:R 600a
- Larm:Visual and audible
- Avfrostning:Automatic
- Display:Monochrome display, touch buttons, external digital
- Dörrlås:Mechanical
- Dörrtyp:Steel, self closing
- Hyllor/lådor:4 adjustable, wire shelves plastic-coated
- Vikt:87 (gross)/78 (net) kg
- Energiklass:C
- Isolering:45/57 mm
- Förp.:1
- B×D×H, inv:620×531×1460 mm
- Temperatur:1...15 °C (ambient 10…40 °C)
Specifications
About this item
Refrigerators designed for professional use. Reliably store non temperature-sensitive items even at ambient temperatures up to +40 °C with a constant cooling power. They are enormously robust, long-lasting, easy to clean and particularly sustainable, not only thanks to natural refrigerant. At the same time, they fulfil all the valid directives, standards and norms, including in terms of hygiene, HACCP concept and GN containers. The efficient cooling system not only ensures low energy and overall operating cost, also safe storage of items not requiring validation of temperature stability and homogeneity.
- Temperature range of +1 to +15 °C, Class M1 with 3 humidity settings
- Forced-air cooling system
- Power failure alarm
- LAN/WiFi optional (SmartModule) and Local AP optional
- 75 cm wide models: GN 2/1, 650×530 mm (crosswise insertion)
- Internal liner material made of commercial-grade moulded polystyrol
Appliances with solid door: Climate class 5, self-closing door at <90° and mechanical lock, aluminium door handle.
Features: Individually adjustable maintenance timer. Castors/adjustable feet as accessory, 75 cm wide appliances with adjustable feet as standard. Steel/white external cabinet finish.
Climate class (energy directive): 5, Climate class (safety standard): 5, Temperature class (energy directive): M1.
75 cm wide models: GN 2/1, 650 × 530 mm (650 mm wide).
Height of adjustable feet: 150 to 180 mm.