Elastic adjustable wrist straps, Jewel®
Katalog # 115-3068
Leverantör: DESCO
Specifikationer
- Beskrivning:Elastic adjustable wrist band, sapphire, 10 mm
- Förp.:1
Specifikationer
Om denna produkt
High visibility bright colour allows supervisor to see that worker is wearing wrist strap.
- Adjustable
- Locking clasp, band locked so it will not slip
- Extra wide, soft elastic band for increased wearer comfort
- Non-shedding, non-sloughing band material, safe for microelectronics
- Nickel-plated brass wristband stud machined to exacting tolerances and precise profile
- Silver impregnated polypropylene knitted on inside for superior conductivity
Replacement elastic bands available; and easy to replace without tools. Unique adjustment design eliminates exposed conductive 'tail' of band and provides added safety eliminating the chance of providing inappropriate ground path.
Medical grade 300 series stainless steel used for all metal parts which eliminates allergic skin reactions for most people.
Certifications: Coil cords are UL listed.
Ordering information: Manufacturer's lifetime warranty on wrist strap stud.