Dual-wire thermoplastic wrist strap
Katalog # 115-3067
Leverantör: DESCO
Specifikationer
- Färg:Blå
- Beskrivning:Dual wire adjustable thermoplastic wrist strap
- Förp.:1
Specifikationer
Om denna produkt
The Dual-Wire thermoplastic wrist strap consists of an insulative exterior with a conductive interior and a 3,4 mm dual conductor jack. The wrist strap is lightweight, low profile, and safely makes contact with human skin.
- Even if one dual-wire wrist strap conductor is severed, operator has reliable path-to-ground with other wire
- Zipper style latching mechanism easily adjusts to most wrist sizes
Made to be used in conjunction with all SCS Workstation Monitors.
Certifications: Meets requirements of EN 61340-5-1 and Wrist Strap standard IEC 61340-4-6.
Ordering information: Designed for use with SCS Dual-Wire Coil Cords (sold separately).