Chemical resistant gloves, AlphaTec® 53-001
Leverantör: Ansell
Certifikat
Om denna produkt
Chemical-resistant gloves with a unique multi-layer design, for broader protection from acids and hydrocarbons.
- Innovative multi-layer polymer design (nitrile/neoprene/nitrile layers)
- Chemical protection against a wide range of chemicals, from acids and bases to hydrocarbons and organic solvents
- Tested according to EN ISO 374:2016, and permeation time of 30 minutes against 13 chemicals out of 18 listed
- Features the latest MICROCHEM chemical barrier technology, providing superior protection for use in hazardous environments
- ANSELL GRIP Technology for handling wet or oily parts with enhanced dexterity, grip, and comfort
- Soft inner nylon liner for comfort and increased mechanical protection
- Oeko-Tex Standard 100 approved
These acid-resistant gloves offer a nitrile and neoprene barrier that provides advanced chemical protection from acids to hydrocarbons and beyond. These AlphaTec 53-001 series black chemical resistant gloves offer Type A chemical protection under EN ISO 374:2016. MICROCHEM Chemical Barrier Technology combines polymers in proprietary blends or multi-layer structures specific to their application, providing superior protection to wearers.
Certifications: EN ISO 374-1:2016 Type A (ACFGJKLMNOPST), EN ISO 374-5:2016, EN 388:2016 (3121A), EN 407 (X1XXXX)
Specifikationer
- Catalog No:
- ANSE53-001/11
- ANSE53-001/10
- ANSE53-001/8
- ANSE53-001/9
- ANSE53-001/6
- ANSE53-001/7
- Ej handspecifika/handspecifika:
- Handspecifika
- Handspecifika
- Handspecifika
- Handspecifika
- Handspecifika
- Handspecifika
- Engångs/Återanvändbar:
- Återanvändbar
- Återanvändbar
- Återanvändbar
- Återanvändbar
- Återanvändbar
- Återanvändbar
- Latex/latexfri:
- Latexfria
- Latexfria
- Latexfria
- Latexfria
- Latexfria
- Latexfria
- Pudrad/puderfri:
- Puderfria
- Puderfria
- Puderfria
- Puderfria
- Puderfria
- Puderfria
- Fodermaterial:
- Nylon
- Nylon
- Nylon
- Nylon
- Nylon
- Nylon
- Yta:
- ANSELL GRIP™ nitrile coating
- ANSELL GRIP™ nitrile coating
- ANSELL GRIP™ nitrile coating
- ANSELL GRIP™ nitrile coating
- ANSELL GRIP™ nitrile coating
- ANSELL GRIP™ nitrile coating
- Kragmodell:
- Kraghandske
- Kraghandske
- Kraghandske
- Kraghandske
- Kraghandske
- Kraghandske
- Handflata, tjocklek:
- 0,84 mm
- 0,84 mm
- 0,84 mm
- 0,84 mm
- 0,84 mm
- 0,84 mm
- EN standard:
- EN ISO 374-1:2016 Type A (ACFGJKLMNOPST), EN ISO 374-5:2016, EN 388:2016 (3121A), EN 407 (X1XXXX)
- EN ISO 374-1:2016 Type A (ACFGJKLMNOPST), EN ISO 374-5:2016, EN 388:2016 (3121A), EN 407 (X1XXXX)
- EN ISO 374-1:2016 Type A (ACFGJKLMNOPST), EN ISO 374-5:2016, EN 388:2016 (3121A), EN 407 (X1XXXX)
- EN ISO 374-1:2016 Type A (ACFGJKLMNOPST), EN ISO 374-5:2016, EN 388:2016 (3121A), EN 407 (X1XXXX)
- EN ISO 374-1:2016 Type A (ACFGJKLMNOPST), EN ISO 374-5:2016, EN 388:2016 (3121A), EN 407 (X1XXXX)
- EN ISO 374-1:2016 Type A (ACFGJKLMNOPST), EN ISO 374-5:2016, EN 388:2016 (3121A), EN 407 (X1XXXX)
- Sterility:
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Antistatisk:
- +
- +
- +
- +
- +
- +