Industrial natural rubber latex gloves, AlphaTec® 87-245 series
Supplier: Ansell
Certificates
About this item
Lightweight natural rubber glove with flocked lining.
- Light-duty glove
- Beaded cuff for easy donning
- Food contact approved
- 100% cotton flock lining
- Good dexterity and tactility
- AQL 1.5
Gloves feature 100% cotton flock for good comfort, and a diamond grip finish on palm and fingers for good dry grip. Tear-resistant beaded cuff for easy donning.
Specifications
- Catalog No:
- ANSE87-245/8.5
- ANSE87-245/9.5
- ANSE87-245/7.5
- ANSE87-245/6.5
- Ej handspecifika/handspecifika:
- Handspecifika
- Handspecifika
- Handspecifika
- Handspecifika
- Engångs/Återanvändbar:
- Återanvändbar
- Återanvändbar
- Återanvändbar
- Återanvändbar
- Latex/latexfri:
- Latex
- Latex
- Latex
- Latex
- Pudrad/puderfri:
- Puderfria
- Puderfria
- Puderfria
- Puderfria
- Material:
- Natural rubber latex
- Natural rubber latex
- Natural rubber latex
- Natural rubber latex
- Kragmodell:
- Med rullad kant
- Med rullad kant
- Med rullad kant
- Med rullad kant
- Handflata, tjocklek:
- 0,43 mm
- 0,43 mm
- 0,43 mm
- 0,43 mm
- Sterility:
- Non sterile
- Non sterile
- Non sterile
- Non sterile