Industrial ESD gloves, EDGE® 48-140
Supplier: Ansell
Certificates
About this item
Multipurpose, light-duty work gloves, combining ESD compatibility, comfort and dexterity.
- Breathable comfort, optimized dexterity
- Seamless polyester and carbon fiber liner provides lightweight comfort
- Anti-static capabilities: Designed to protect workers and electronic products and equipment from the potential dangers of static electricity
- Can be used in explosion hazard (ATEX) zones as part of a grounding chain
The EDGE 48-140 series glove is made of a seamless 13 gauge polyester and carbon fiber liner, with polyurethane palm coating. Electrostatic discharge product protection is ensured due to the presence of carbon fiber. The coated palm also prevents the transfer of perspiration to electronic parts. The 48-140 series is the practical choice for light applications involving assembly and handling tasks in electronic/electrical industries. Gloves meet the requirements for electrostatic dissipative materials per EN 1149-1/2/3.
Certifications: EN 388:2016 (3121A), EN 16350; Cat. II
Specifications
- Catalog No:
- ANSE48-140/10
- ANSE48-140/11
- ANSE48-140/8
- ANSE48-140/9
- ANSE48-140/6
- ANSE48-140/7
- Ej handspecifika/handspecifika:
- Handspecifika
- Handspecifika
- Handspecifika
- Handspecifika
- Handspecifika
- Handspecifika
- Engångs/Återanvändbar:
- Återanvändbar
- Återanvändbar
- Återanvändbar
- Återanvändbar
- Återanvändbar
- Återanvändbar
- Latex/latexfri:
- Latexfria
- Latexfria
- Latexfria
- Latexfria
- Latexfria
- Latexfria
- Material:
- Polyester/kol
- Polyester/kol
- Polyester/kol
- Polyester/kol
- Polyester/kol
- Polyester/kol
- Yta:
- Palm PU dipped
- Palm PU dipped
- Palm PU dipped
- Palm PU dipped
- Palm PU dipped
- Palm PU dipped
- Mätare:
- 13
- 13
- 13
- 13
- 13
- 13
- Kragmodell:
- Stickad mudd
- Stickad mudd
- Stickad mudd
- Stickad mudd
- Stickad mudd
- Stickad mudd
- EN standard:
- EN 388:2016 (3121A), EN 16350
- EN 388:2016 (3121A), EN 16350
- EN 388:2016 (3121A), EN 16350
- EN 388:2016 (3121A), EN 16350
- EN 388:2016 (3121A), EN 16350
- EN 388:2016 (3121A), EN 16350
- Sterility:
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Antistatisk:
- +
- +
- +
- +
- +
- +