Cut resistant gloves, HyFlex® 11-730
Supplier: Ansell
Certificates
About this item
Durable polyurethane gloves, offering advanced cut protection and confident grip.
- PU dipping enhances grip performance, making the gloves ideal for medium-duty handling of dry or slightly oily components
- INTERCEPT™ Cut Resistance Technology yarns and fibers help ensure advanced, EN ISO D cut protection levels
- PU formulation enhances durability, facilitating repeated use over extended periods
- First-rate liner composition, made up of quality materials, including Kevlar® and nylon
- Knitwrist cuff ensures hand and wrist protection with optimal comfort
The glove liner provides good levels of cut resistance for safe use in controlling, handling, and assembly applications involving contact with sharp pieces. The specific formulation of the polyurethane dipping provides a secure grip on dry or slightly oily pieces.
Recommended for: Assembling metal parts and components; assembly of white goods; metalwork, glass processing; handling sharp or irregular metal pieces; press injection molding.
Certifications: EN 388:2016 (4X43D), EN 407 (X1XXXX); Cat. II
Specifications
- Catalog No:
- ANSE11-730/11
- ANSE11-730/10
- ANSE11-730/9
- ANSE11-730/7
- ANSE11-730/8
- ANSE11-730/6
- Ej handspecifika/handspecifika:
- Handspecifika
- Handspecifika
- Handspecifika
- Handspecifika
- Handspecifika
- Handspecifika
- Engångs/Återanvändbar:
- Återanvändbar
- Återanvändbar
- Återanvändbar
- Återanvändbar
- Återanvändbar
- Återanvändbar
- Yta:
- Palm PU dipped
- Palm PU dipped
- Palm PU dipped
- Palm PU dipped
- Palm PU dipped
- Palm PU dipped
- Mätare:
- 13
- 13
- 13
- 13
- 13
- 13
- Kragmodell:
- Stickad mudd
- Stickad mudd
- Stickad mudd
- Stickad mudd
- Stickad mudd
- Stickad mudd
- EN standard:
- EN 388:2016 (4X43D), EN 407 (X1XXXX)
- EN 388:2016 (4X43D), EN 407 (X1XXXX)
- EN 388:2016 (4X43D), EN 407 (X1XXXX)
- EN 388:2016 (4X43D), EN 407 (X1XXXX)
- EN 388:2016 (4X43D), EN 407 (X1XXXX)
- EN 388:2016 (4X43D), EN 407 (X1XXXX)