VWR®, Caps for Erlenmeyer Shaker Flasks
Leverantör: VWR Collection
VWR®, Caps for Erlenmeyer Shaker Flasks
Leverantör: VWR Collection
Certificates
Om denna produkt
12 Alternativ tillgängliga nedan
Product Details & Documents
CoC_VWRI214-0482_2404071769F COC_VWRI214-0487_230216771F coc_VWRI214-0491_2510251908A coc_VWRI214-0492_2511081909A coc_VWRI214-0493_2510251909A
More
Specifications for Product Family
Specifikationer
- Catalog No:
- 214-0482
- 214-0483
- 214-0484
- 214-0485
- 214-0486
- 214-0487
- 214-0488
- 214-0489
- 214-0490
- 214-0491
- 214-0492
- 214-0493
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Miljöcertificering:
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
Specifikationer
Product Family Options
Produktinformation
- TillgänglighetPrice
- 214-0482Plug seal cap for 125 mlSpecifikationer:
Low Manufacturing Impact More
Specifications
Cat. No.214-0482Environmentally PreferableLow Manufacturing ImpactMiljöcertificeringISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - 214-0483Vent cap for 125 mlSpecifikationer:
Low Manufacturing Impact More
Specifications
Cat. No.214-0483Environmentally PreferableLow Manufacturing ImpactMiljöcertificeringISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - 214-0484Plug seal cap for 250 mlSpecifikationer:
Low Manufacturing Impact More
Specifications
Cat. No.214-0484Environmentally PreferableLow Manufacturing ImpactMiljöcertificeringISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - 214-0485Vent cap for 250 mlSpecifikationer:
Low Manufacturing Impact More
Specifications
Cat. No.214-0485Environmentally PreferableLow Manufacturing ImpactMiljöcertificeringISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - 214-0486Plug seal cap for 500 ml, 1000 mlPlug seal cap for 500 ml, 1000 mlLog in för att kolla tillgänglighet och ert pris1 * 25 Items305,00 krSpecifikationer:
Low Manufacturing Impact More
Specifications
Cat. No.214-0486Environmentally PreferableLow Manufacturing ImpactMiljöcertificeringISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - 214-0487Vent cap for 500 ml, 1000 mlSpecifikationer:
Low Manufacturing Impact More
Specifications
Cat. No.214-0487Environmentally PreferableLow Manufacturing ImpactMiljöcertificeringISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - 214-0488Plug seal cap for 2000 mlSpecifikationer:
Low Manufacturing Impact More
Specifications
Cat. No.214-0488Environmentally PreferableLow Manufacturing ImpactMiljöcertificeringISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - 214-0489Vent cap for 2000 mlSpecifikationer:
Low Manufacturing Impact More
Specifications
Cat. No.214-0489Environmentally PreferableLow Manufacturing ImpactMiljöcertificeringISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - 214-0490Plug seal cap for 3000 mlSpecifikationer:
Low Manufacturing Impact More
Specifications
Cat. No.214-0490Environmentally PreferableLow Manufacturing ImpactMiljöcertificeringISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - 214-0491Vent cap for 3000 mlSpecifikationer:
Low Manufacturing Impact More
Specifications
Cat. No.214-0491Environmentally PreferableLow Manufacturing ImpactMiljöcertificeringISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - 214-0492Plug seal cap for 5000 mlSpecifikationer:
Low Manufacturing Impact More
Specifications
Cat. No.214-0492Environmentally PreferableLow Manufacturing ImpactMiljöcertificeringISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - 214-0493Vent cap for 5000 mlSpecifikationer:
Low Manufacturing Impact More
Specifications
Cat. No.214-0493Environmentally PreferableLow Manufacturing ImpactMiljöcertificeringISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact