Elbows
Leverantör: LABCONCO
Gå till:
Om denna produkt
PVC elbows 90° are compatible with thermoplastic duct.
Designed and engineered for quick installation and minimum pressure losses, elbows feature belled end connections to receive PVC duct directly.
3 Alternativ tillgängliga nedan
Product Details & Documents
PVC elbows 90° are compatible with thermoplastic duct.
Designed and engineered for quick installation and minimum pressure losses, elbows feature belled end connections to receive PVC duct directly.
Product Family Options
Produktinformation
- TillgänglighetPrice
- LBCP470870090° elbows, Ø 152 mm, Height - 343 mm
- LBCP702730090° elbows, Ø 254 mm, Height - 508 mm
- LBCP560210090° elbows, Ø 305 mm, Height - 610 mm