MIU medium base dehydrated culture media
Katalog # HIMEM1076-500G
Leverantör: HIMEDIA
CAS-nummer:
Specifikationer
- Förp.:500 g
- Beskrivning:MIU medium base
- Seq:01
Specifikationer
Om denna produkt
For the detection of motility and urease and indole production.
- Microbiological culture media
- 500 g poly bottles