L.D. agar dehydrated culture media
HIMEM742-500G
: HIMEDIA
:
- Förp.:500 g
- Beskrivning:L.D. agar
- Seq:01
For the cultivation and identification of fastidious anaerobic bacteria.
- Microbiological culture media
- 500 g poly bottles
For the cultivation and identification of fastidious anaerobic bacteria.