EMB agar dehydrated culture media
Katalog # HIMEM317-500G
Leverantör: HIMEDIA
CAS-nummer:
Specifikationer
- Förp.:500 g
- Beskrivning:EMB agar
Specifikationer
Om denna produkt
For the differential isolation of gram-negative Enteric bacilli from clinical and nonclinical specimens.
- Microbiological culture media
- 500 g poly bottles