Clopyralid Analytical standard, reference material, Fluka™
Katalog # HONCR1814-100MG
Leverantör: Honeywell Chemicals
CAS-nummer:
Specifikationer
- Förpackningstyp:Glass Bottle
- Förp.:100 mg
- Standard:Analytical standard
- CAS-nr:1702-17-6
- Beskrivning:Clopyralid
- Storage conditions:Ambient
- MDL No.:MFCD00078655
- Synonym(s):3,6-Dichloro-pyridine-2-carboxylic acid
- Smältpunkt:151 - 152 °C
- Formel:C₆H₃Cl₂NO₂
- Molekylvikt:192.00 g/mol
- Benämning:Clopyralid
- Fysikalisk form:Crystalline
Specifikationer
Om denna produkt
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery