Specification certified 20 ml screw clear glass vial and 24 mm closed top screw cap kits for scintillation, level 2, SureSTART™
Catalog # HYPE6AK20SCIN
Supplier: Thermo Fisher Scientific
Specifications
- Med lock:Ja
- Lockmaterial:PP
- Lock storlek:24 mm
- Diam×H:28×58 mm
- Septum:Silver PP foil scintillation grade
- Kapacitet:20,00 ml
- Färg:Klar
- Beskrivning:Assembled screw scintillation vial kit
- Material:Glas
- Typ av lock:Closed top
- Lock, färg:Vit
- Botten:Flat bottom
- Förp.:500
Specifications
About this item
Perform accurate liquid scintillation counting while safely containing radioactive cocktails with our level 2 Thermo Scientific™ SureSTART™ specification certified screw vial and cap kits for scintillation.
Select level 2 products for high throughput applications, when robustness and reproducibility are key. Our kits for scintillation include 20 ml glass screw vials and closed top polypropylene caps lined with polypropylene foil septa.