L-Photo-Leucine, Pierce™
Katalog # PIER22610
Leverantör: Thermo Fisher Scientific
CAS-nummer:
Specifikationer
- Förp.:100 mg
- Seq:2440
Specifikationer
Specifikationstestresultat
- Product Line:Pierce™
- Labeling Method:Chemical Labeling, Metabolic Labeling
- Crosslinker Type:Heterobifunctional
- Reactive Moiety:Diazirine
- Spacer:Short
- Form:Solid
- Quantity:100 mg
- Type:Crosslinker
- Solubility:Water
Specifikationstestresultat
Om denna produkt
:
:143.17 g/mol
Förvaringstemperatur:Kylskåp
Formel:C₅H₉O₂N₃