16Alpha-Hydroxy Prednisolone-9(11)-ene, Pharmaceutical Analytical Impurities
1A01260.
: USP
:
- Förpackningstyp:Vial
- Förp.:25 mg
- Standard:Pharmaceutical Analytical Impurities
- CAS-nr:39672-76-9
- Beskrivning:16Alpha-Hydroxy Prednisolone-9(11)-ene
- Synonym(s):(8S,10S,13S,14S,16R,17S)-16,17-dihydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-6,7,8,10,12,13,14,15,16,17-decahydro-3Hcyclopenta[a]phenanthren-3-one
- Benämning:16Alpha-Hydroxy Prednisolone-9(11)-ene
Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.