16Alpha-Hydroxy Prednisolone-14-ene, Pharmaceutical Analytical Impurities
Katalog # 1A01230.
Leverantör: USP
CAS-nummer:
Specifikationer
- Förpackningstyp:Vial
- Förp.:25 mg
- Standard:Pharmaceutical Analytical Impurities
- CAS-nr:131918-73-5
- Beskrivning:16Alpha-Hydroxy Prednisolone-14-ene
- Synonym(s):(8R,9S,10R,11S,13S,16R,17S)-11,16,17-trihydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-6,7,8,9,10,11,12,13,16,17-decahydro-3H-cyclopenta[a]phenanthren-3-one
- Benämning:16Alpha-Hydroxy Prednisolone-14-ene
Specifikationer
Om denna produkt
Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.