Pemetrexed Acid N-Hydroxysuccinimide Ester, Pharmaceutical Analytical Impurities
Catalog # 1A00630.
Supplier: USP
CAS Number:
Specifications
- Förpackningstyp:Vial
- Förp.:25 mg
- Standard:Pharmaceutical Analytical Impurities
- CAS-nr:204257-65-8
- Beskrivning:Pemetrexed Acid N-Hydroxysuccinimide Ester
- Synonym(s):2,5-dioxopyrrolidin-1-yl 4-(2-(2-amino-4-oxo-4,7-dihydro-3H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl)benzoate
- Benämning:Pemetrexed Acid N-Hydroxysuccinimide Ester
Specifications
About this item
Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.