Order Entry
Sweden
ContactUsLinkComponent
Atorvastatin 3-deoxyhept-2-enoic Acid, Pharmaceutical Analytical Impurities
Atorvastatin 3-deoxyhept-2-enoic Acid, Pharmaceutical Analytical Impurities
Katalog # 1A00760.
Supplier:  USP
CAS Number:  
Atorvastatin 3-deoxyhept-2-enoic Acid, Pharmaceutical Analytical Impurities
Katalog # 1A00760.
Supplier:  USP
CAS Number:  

Specifications

  • Förpackningstyp:
    Vial
  • Förp.:
    25 mg
  • Standard:
    Pharmaceutical Analytical Impurities
  • CAS-nr:
    1105067-93-3
  • Beskrivning:
    Atorvastatin 3-deoxyhept-2-enoic Acid
  • Synonym(s):
    (S,E)-7-(2-(4-fluorophenyl)-5-isopropyl-3-phenyl-4-(phenylcarbamoyl)-1H-pyrrol-1-yl)-5-hydroxyhept-2-enoic acid
  • Benämning:
    Atorvastatin 3-deoxyhept-2-enoic Acid

Specifications

About this item

Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.