Tolterodine Dimer, Pharmaceutical Analytical Impurities
Catalog # 1A01620.
Supplier: USP
CAS Number:
Specifications
- Förpackningstyp:Vial
- Förp.:25 mg
- Standard:Pharmaceutical Analytical Impurities
- CAS-nr:854306-72-2
- Beskrivning:Tolterodine Dimer
- Synonym(s):2,2'-((isopropylazanediyl)bis(1-phenylpropane-3,1-diyl))bis(4-methylphenol)
- Benämning:Tolterodine Dimer
Specifications
About this item
Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.