Order Entry
Sweden
ContactUsLinkComponent
Tetramethyl Ethylene Diamine, Pharmaceutical Analytical Impurities
Tetramethyl Ethylene Diamine, Pharmaceutical Analytical Impurities
  1A00400.
 :  USP
 :  
Tetramethyl Ethylene Diamine, Pharmaceutical Analytical Impurities
  1A00400.
 :  USP
 :  

 

  • Förpackningstyp:
    Ampule
  • Förp.:
    25 mg
  • Standard:
    Pharmaceutical Analytical Impurities
  • CAS-nr:
    110-18-9
  • Beskrivning:
    Tetramethyl Ethylene Diamine
  • UN No.:
    2372
  • Synonym(s):
    N1,N1,N2,N2-tetramethylethane-1,2-diamine
  • Benämning:
    Tetramethyl Ethylene Diamine

 

 

Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.