Order Entry
Sweden
ContactUsLinkComponent
TEMED Dihydrochloride, Pharmaceutical Analytical Impurities
TEMED Dihydrochloride, Pharmaceutical Analytical Impurities
Katalog # 1A01290.
Leverantör:  USP
CAS-nummer:  
TEMED Dihydrochloride, Pharmaceutical Analytical Impurities
Katalog # 1A01290.
Leverantör:  USP
CAS-nummer:  

Specifikationer

  • Förpackningstyp:
    Vial
  • Förp.:
    25 mg
  • Standard:
    Pharmaceutical Analytical Impurities
  • CAS-nr:
    7677-21-6
  • Beskrivning:
    TEMED Dihydrochloride
  • Synonym(s):
    N1,N1,N2,N2-tetramethylethane-1,2-diamine, hydrochloride
  • Benämning:
    TEMED Dihydrochloride

Specifikationer

Om denna produkt

Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.