Rosuvastatin Isoamyl Ester, Pharmaceutical Analytical Impurities
Katalog # 1A00170.
Leverantör: USP
CAS-nummer:
Specifikationer
- Förpackningstyp:Vial
- Förp.:25 mg
- Standard:Pharmaceutical Analytical Impurities
- CAS-nr:1197348-98-3
- Beskrivning:Rosuvastatin Isoamyl Ester
- Synonym(s):(3S,5R,6E)-7-[4-(4-flurophenyl)-6-(1-methylethyl)-2-[N-methyl(methylsulphonyl)amino]-5-pyrimidinyI]-3,5-dihydroxy-6-heptanoic acid, methyl butyl ester
- Benämning:Rosuvastatin Isoamyl Ester
Specifikationer
Om denna produkt
Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.