Rivaroxaban Glycolic Acid Analog, Pharmaceutical Analytical Impurities
1A01170.
: USP
:
- Förp.:25 mg
- Standard:Pharmaceutical Analytical Impurities
- CAS-nr:931204-39-6
- Beskrivning:Rivaroxaban Glycolic Acid Analog
- Synonym(s):(S)-2-(2-((4-(5-((5-chlorothiophene-2-carboxamido)methyl)-2-oxooxazolidin-3-yl)phenyl)amino)ethoxy)acetic acid
- Benämning:Rivaroxaban Glycolic Acid Analog
Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.