Risperidone Difluoroketone, Pharmaceutical Analytical Impurities
1A00600.
: USP
:
- Förpackningstyp:Vial
- Förp.:10 mg
- Standard:Pharmaceutical Analytical Impurities
- CAS-nr:158697-67-7
- Beskrivning:Risperidone Difluoroketone
- Synonym(s):3-(2-(4-(2,4-difluorobenzoyl)piperidin-1-yl)ethyl)-2-methyl-6,7,8,9-tetrahydro-4H-pyrido[1,2-a]pyrimidin-4-one
- Benämning:Risperidone Difluoroketone
Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.