Order Entry
Sweden
ContactUsLinkComponent
Pyrrolic acid Derivative, Pharmaceutical Analytical Impurities
Pyrrolic acid Derivative, Pharmaceutical Analytical Impurities
Katalog # 1A00460.
Leverantör:  USP
CAS-nummer:  
Pyrrolic acid Derivative, Pharmaceutical Analytical Impurities
Katalog # 1A00460.
Leverantör:  USP
CAS-nummer:  

Specifikationer

  • Förpackningstyp:
    Vial
  • Förp.:
    10 mg
  • Standard:
    Pharmaceutical Analytical Impurities
  • CAS-nr:
    404839-11-8
  • Beskrivning:
    Pyrrolic acid Derivative
  • Synonym(s):
    4-(2-hydroxyethyl)-1H-pyrrole-3-carboxylic acid
  • Benämning:
    Pyrrolic acid Derivative

Specifikationer

Om denna produkt

Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.