Orders LinkContactUsLinkComponent
Paliperidone Resolution Mixture, Reference Standard
Paliperidone Resolution Mixture, Reference Standard
Paliperidone Resolution Mixture, USP reference standard, Vial, 20 mg
Katalog # 1491795.
Leverantör:   USP
Paliperidone Resolution Mixture, Reference Standard
Katalog # 1491795.
Leverantör:  USP
CAS-nummer:  
Paliperidone Resolution Mixture, Reference Standard
Paliperidone Resolution Mixture, USP reference standard, Vial, 20 mg
CAS-nummer:  
Pris
15 350,00 kr
Log in för tillgänglighet
1 * 20 mg
Produktdetaljer och dokument
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Specifikationer
Specifikationer
  • Förpackningstyp:
    Vial
  • Förp.:
    20 mg
  • Standard:
    USP reference standard
  • Beskrivning:
    Paliperidone Resolution Mixture
  • UN No.:
    2811
  • Cat. No.:
    1491795.
  • Benämning:
    Paliperidone Resolution Mixture
Specifikationer
Rekommendationer
Rekommendationer anpassas endast baserat på dina shoppingpreferenser om du har gett ditt samtycke genom att aktivera "Förbättra min shoppingupplevelse" på sidan "Personlig information".
Annars får du generiska rekommendationer.