Order Entry
Sweden
ContactUsLinkComponent
N-Ethyl Quetiapine, Pharmaceutical Analytical Impurities
N-Ethyl Quetiapine, Pharmaceutical Analytical Impurities
Katalog # 1A00180.
Leverantör:  USP
CAS-nummer:  
N-Ethyl Quetiapine, Pharmaceutical Analytical Impurities
Katalog # 1A00180.
Leverantör:  USP
CAS-nummer:  

Specifikationer

  • Förpackningstyp:
    Vial
  • Förp.:
    25 mg
  • Standard:
    Pharmaceutical Analytical Impurities
  • CAS-nr:
    1011758-03-4 (Free base)
  • Beskrivning:
    N-Ethyl Quetiapine
  • Synonym(s):
    11-(4-Ethylpiperazin-1-yl)dibenzo[b,f][1,4]thiazepine hydrochloride
  • Benämning:
    N-Ethyl Quetiapine

Specifikationer

Om denna produkt

Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.