Order Entry
Sweden
ContactUsLinkComponent
 
N-Benzylacetamide, Pharmaceutical Analytical Impurities
  1A01810.
 :  USP
 :  
N-Benzylacetamide, Pharmaceutical Analytical Impurities
  1A01810.
 :  USP
 :  

 

  • Förpackningstyp:
    Vial
  • Förp.:
    25 mg
  • Standard:
    Pharmaceutical Analytical Impurities
  • CAS-nr:
    588-46-5
  • Beskrivning:
    N-Benzylacetamide
  • Synonym(s):
    N-Acetylbenzylamine
  • Benämning:
    N-Benzylacetamide

 

 

Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.