Orders LinkContactUsLinkComponent
Cellulose, Reference Standards
Cellulose, Reference Standards
Leverantör:   USP
Cellulose, Reference Standards
Leverantör:  USP
Certifikat
2 Alternativ tillgängliga nedan
Produktdetaljer och dokument
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Dokument
Specifikationer för produkt
Specifikationer
Specifikationer
  • Catalog No:
  • 1098388.
  • 1098399.
  • Benämning:
  • Microcrystalline Cellulose
  • CAS-nr:
  • 9004-34-6
Specifikationer
Product Family Options
Produktinformation
  • Standard
    Förpackningstyp
    Förp.
    Tillgänglighet
    Price
  • 1098388.
    Microcrystalline Cellulose
    Microcrystalline Cellulose
    USP reference standard
    Vial
    1 g
    Log in för tillgänglighet
    Log in för tillgänglighet
    1 * 1 g
    1 * 1 g - 4 900,00 kr
    4 900,00 kr
    1 * 1 g
       4 900,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    1098388.
    Benämning
    Microcrystalline Cellulose
    CAS-nr
    9004-34-6
  • 1098399.
    Silicified Microcrystalline Cellulose
    Silicified Microcrystalline Cellulose
    USP reference standard
    Vial
    1 g
    Log in för tillgänglighet
    Log in för tillgänglighet
    1 * 1 g
    1 * 1 g - 5 400,00 kr
    5 400,00 kr
    1 * 1 g
       5 400,00 kr
    Specifikationer:
    More
    Specifications
    Cat. No.
    1098399.
    Benämning
    CAS-nr