Fluoroandrostadiene Carboxylic Acid, Pharmaceutical Analytical Impurities
1A00120.
: USP
:
- Förpackningstyp:Vial
- Förp.:25 mg
- Standard:Pharmaceutical Analytical Impurities
- CAS-nr:37927-01-8
- Beskrivning:Fluoroandrostadiene Carboxylic Acid
- Synonym(s):(8S,9R,10S,11S,13S,14S,16R,17R)-9-fluoro-11,17-dihydroxy-10,13,16-trimethyl-3-oxo-6,7,8,9,10,11,12,13,14,15,16,17-dodecahydro-3H-cyclopenta[a]phenanthrene-17-carboxylic acid
- Benämning:Fluoroandrostadiene Carboxylic Acid
Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.