Dimethylethylamine Hydrochloride, Pharmaceutical Analytical Impurities
1A01320.
: USP
:
- Förpackningstyp:Vial
- Förp.:25 mg
- Standard:Pharmaceutical Analytical Impurities
- CAS-nr:58114-25-3
- Beskrivning:Dimethylethylamine Hydrochloride
- Synonym(s):N,N-dimethylethanamine, hydrochloride
- Benämning:Dimethylethylamine Hydrochloride
Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.