Order Entry
Sweden
ContactUsLinkComponent
Dichlorothiophene Carboxylic Acid, Pharmaceutical Analytical Impurities
Dichlorothiophene Carboxylic Acid, Pharmaceutical Analytical Impurities
Catalog # 1A01580.
Supplier:  USP
CAS Number:  
Dichlorothiophene Carboxylic Acid, Pharmaceutical Analytical Impurities
Catalog # 1A01580.
Supplier:  USP
CAS Number:  

Specifications

  • Förpackningstyp:
    Vial
  • Förp.:
    25 mg
  • Standard:
    Pharmaceutical Analytical Impurities
  • CAS-nr:
    89166-94-9
  • Beskrivning:
    Dichlorothiophene Carboxylic Acid
  • Synonym(s):
    3,5-dichlorothiophene-2-carboxylic acid
  • Benämning:
    Dichlorothiophene Carboxylic Acid

Specifications

About this item

Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.