Desfluorocitalopram Oxalate, Pharmaceutical Analytical Impurities
Katalog # 1A01750.
Supplier: USP
CAS Number:
Specifications
- Förpackningstyp:Vial
- Förp.:25 mg
- Standard:Pharmaceutical Analytical Impurities
- CAS-nr:1093072-86-6
- Beskrivning:Desfluorocitalopram Oxalate
- Synonym(s):1-(3-(dimethylamino)propyl)-1-phenyl-1,3-dihydroisobenzofuran-5-carbonitrile, oxalate
- Benämning:Desfluorocitalopram Oxalate
Specifications
About this item
Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.