Decarboxy Levofloxacin, Pharmaceutical Analytical Impurities
Catalog # 1A00160.
Supplier: USP
CAS Number:
Specifications
- Förpackningstyp:Vial
- Förp.:25 mg
- Standard:Pharmaceutical Analytical Impurities
- CAS-nr:178964-53-9
- Beskrivning:Decarboxy Levofloxacin
- Synonym(s):(S)-9-fluoro-3-methyl-7-oxo-10-(4-methylpiperazine-1-yl)-2,3-dihydro-7H-pyrido [1,2,3-de] -1,4-benzoxazine
- Benämning:Decarboxy Levofloxacin
Specifications
About this item
Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.