Order Entry
Sweden
ContactUsLinkComponent
Budesonide Pyruvic Acid, Pharmaceutical Analytical Impurities
Budesonide Pyruvic Acid, Pharmaceutical Analytical Impurities
Katalog # 1A02000.
Leverantör:  USP
CAS-nummer:  
Budesonide Pyruvic Acid, Pharmaceutical Analytical Impurities
Katalog # 1A02000.
Leverantör:  USP
CAS-nummer:  

Specifikationer

  • Förpackningstyp:
    Vial
  • Förp.:
    25 mg
  • Standard:
    Pharmaceutical Analytical Impurities
  • Beskrivning:
    Budesonide Pyruvic Acid
  • Synonym(s):
    2-((6aR,6bS,7S,8aS,8bS,11aR,12aS,12bS)-7-hydroxy-6a,8a-dimethyl-4-oxo-10-propyl-1,2,4,6a,6b,7,8,8a,11a,12,12a,12b-dodecahydro-8bH-naphtho[2',1':4,5]indeno[1,2-d][1,3]dioxol-8b-yl)-2-oxoacetic acid
  • Benämning:
    Budesonide Pyruvic Acid

Specifikationer

Om denna produkt

Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.