Order Entry
Sweden
ContactUsLinkComponent
Budesonide Acetaldehyde Acetal, Pharmaceutical Analytical Impurities
Budesonide Acetaldehyde Acetal, Pharmaceutical Analytical Impurities
Katalog # 1A00510.
Leverantör:  USP
CAS-nummer:  
Budesonide Acetaldehyde Acetal, Pharmaceutical Analytical Impurities
Katalog # 1A00510.
Leverantör:  USP
CAS-nummer:  

Specifikationer

  • Förpackningstyp:
    Vial
  • Förp.:
    25 mg
  • Standard:
    Pharmaceutical Analytical Impurities
  • CAS-nr:
    1040085-98-0
  • Beskrivning:
    Budesonide Acetaldehyde Acetal
  • Synonym(s):
    16α,17-[(1RS)-ethylidenebis(oxy)]-11β,21-dihydroxypregna-1,4-diene-3,20-dione
  • Benämning:
    Budesonide Acetaldehyde Acetal

Specifikationer

Om denna produkt

Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.