Bis(dimethylaminoethyl) Ether Dihydrochloride, Pharmaceutical Analytical Impurities
Katalog # 1A01300.
Leverantör: USP
CAS-nummer:
Specifikationer
- Förpackningstyp:Vial
- Förp.:25 mg
- Standard:Pharmaceutical Analytical Impurities
- CAS-nr:103526-47-2
- Beskrivning:Bis(dimethylaminoethyl) Ether Dihydrochloride
- Synonym(s):2,2'-oxybis(N,N-dimethylethan-1-amine), hydrochloride
- Benämning:Bis(dimethylaminoethyl) Ether Dihydrochloride
Specifikationer
Om denna produkt
Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.